11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific
Proveedor: Thermo Scientific
C4011-72AW C4011-V1 C4011-6W C4011-9TR C4011-73W C4013-17 C4011-73P C4011-87AW C4011-9 C4011-V5 C4011-1Y C4011-S2W C4011-87 60180-679 C4011-88W C4011-92 C4013-15P C4011-LV1 C4011-72W C4011-V6 C4011-87W C4011-1W C4011-LV2 C4011-S4 C4011-S5W C4011-73 C4011-89W C4011-S6W C4011-76PPS C4011-74 C4011-4 C4011-1 C4011-LV1W C4011-S1W C4011-88 C4011-1G C4011-1R C4011-1B C4011-S1 C4011-72P C4013-S15 C4011-95 C4011-5W C4011-2W C4011-LV2W C4011-6 C4011-5 C4011-74W C4011-S5
66030-554CS 15407.66 MXN
66064-252 66064-210 66064-254 66064-256 66064-212 66064-216 66030-452 66064-218 66011-241 66030-170 66030-174 66030-130 66064-250 89235-950 66064-246 89235-952 66064-248 66064-920 66064-208 66064-924 66064-926 66064-918 66030-184 89235-904 97054-222 89235-962 66064-230 89235-960 66030-550 66030-556 66064-958 66064-916 66030-554 66011-181 76202-158 66064-194 66021-192 66064-222 66064-228 66030-168 66030-124 66030-128 66030-126 66065-260 66030-160 66030-120 69400-092 66030-162 66064-260
11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific
Viales
Use Thermo Scientific 11 mm Crimp/Snap Top Vials with snap caps or aluminum crimp seal closures. These high-quality 11 mm glass vials are 2 ml, 12×32 mm and fit most brands of autosamplers. Made of superior quality 33 expansion borosilicate clear Type 1, Class A or 51A Type 1, Class B amber glass, they meet all requirements of US, EU, and JPN Pharmacopia.
Easy-to-fill wide-neck opening vials require 6 mm diameter micro-inserts.
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific 11 mm Crimp Top Clear Vial Convenience Kits (Unassembled). Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures, while keeping matching supplies together. Clear trays make it easy to keep track of available supplies without opening containers.
Identify sample and volume with graduated, write-on patch provided on certain vials. Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Obtain optimal recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). Polyspring inserts are self-aligning and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Pulled point inserts are an economical choice for noncritical applications. Compatible with most HPLC and GC autosamplers.