HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Proveedor: Air-Tite
HENKE-JECT®
5100.000V0 5200.000V0 8300018745 5300.000D0 5300.X00D0 5100.X00V0 8300014579 5050.X00V0 8300005762 8300006682 5050.000VZ 5200.X00V0
89215-236CS 15622.85 MXN
10148-330 89215-217 89215-218 89215-240 89215-230 89215-232 89215-222 89215-234 89215-225 89215-236 89215-226 89215-238
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Jeringas Jeringas de uso múltiple
HENKE-JECT® syringes are a high quality disposable rubber-piston syringe offering substantial savings.
  • ETO gas sterilized
  • Latex and pyrogen free
A lubricated synthetic rubber piston allows for a smooth draw while a positive stop helps prevent accidental pullout. Bold graduations are easily read and resist wipe off.
Certificaciones: Meets ISO standards, CE registered.
Envase: All HENKE-JECT® syringes are packaged within individual sterile strip packs.