Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
Proveedor: Honeywell Research Chemicals
HYDRANAL™
34868-6X500ML-US 34829-1L 34868-6X100ML 34868-6X500ML-US 34829-6X1L 34821-6X50ML 34836-6X500ML-US 34868-100ML 34868-500ML-US 34820-6X500ML-US 34807-6X500ML-US 34840-50ML-R 34840-6X50ML-R 34840-6X50ML 34843-6X500ML-US 34836-6X1L-US 34739-6X500ML-US 34726-500ML 34810-6X500ML-US 34810-6X500ML-US 34843-6X500ML-US 34820-6X500ML-US
BJ34821-50MLCS 59313.04 MXN
BJ34868500MLUS 220059-070 BJ34821-50ML 220059-071 BJ34868-500ML BJ34810500MLUS BJ34820500MLUS BJ34840-50ML BJ34843-500ML BJ34843500MLUS BJ34810-500ML BJ34820-500ML BJ34829-1L BJ34836500MLUS BJ34739500MLUS BJ34836-1L-US BJ34829-6X1L BJ34868-100ML BJ34868-500MLS BJ34868-6100ML BJ34807500MLUS BJ34726-500ML
Hydranal® Coulometric Karl Fischer Titration Reagents, Honeywell Research Chemicals
Karl Fischer titulantes y reactivos
Coulometric Karl Fischer titrations normally require two reagent solutions: an anolyte (the solution in the anodic compartment) and a catholyte (the solution in the cathodic compartment).
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range available
  • Long shelf life (up to five years)