VWR® Mason's Form Hygrometer
61503-0411
82020-835CS 3585.09 MXN
82020-835
VWR® Mason's Form Hygrometer
Displays relative humidity and dew point using wet and dry bulb thermometers.
  • Clear styrene reservoir
  • White back glass type
Two organic liquid filled thermometers are secured in a sturdy styrene housing. A clear styrene reservoir and wick supplies water to the wet bulb. Complete with laminated humidity chart and hole for wall mounting. Organic thermometer fill.