VWR® Volumetric Flask with PE Stopper, Class A
Proveedor: VWR
VWR®
134.232.07 134.232.01 134.232.08 134.232.06 131.232.10 131.232.09 134.232.02 134.232.04 131.232.11 134.232.05
10545-990CS 5342.2 MXN
10545-990 10545-992 10545-994 10545-984 10545-996 10545-986 10545-998 10545-988 10546-002 10546-000
VWR® Volumetric Flask with PE Stopper, Class A
Matraces Matraces aforados
These volumetric flasks are batch certified and are calibrated at 20 degrees Celsius To Contain.
  • Borosilicate glass fabrication
  • Polyethylene stopper construction
  • Durable white marking spot
They feature a single graduation ring and polyethylene stopper with marking spot.
Made from ASTM E438 Type 1 Class A Borosilicate 33 expansion glass as per ASTM specification E237 and E288. Durable white color printing which has good life in laboratory environment.