SpillStopper Work Surfaces, Labconco
9501420 9501610 9501402 9501500 9501311 9501621 9501511 9501612 9501510 9501512 9501312 9501801 9501421 9501412 9501322 9501611 9501522 9501320 9501602 9501620 9501600 9501822 9501401 9501321 9501811 9501410 9501300 9501301 9501422 9501411 9501802 9501520 9501601 9501502 9501400 9501821 9501521 9501800 9501310 9500300 9501501 9501302 9501812 9501810 9501622 9501820
10018-928EA 21700.87 MXN
10769-448 10769-404 10769-446 10769-402 10769-468 10769-488 10769-444 10769-400 10769-466 10769-442 10769-464 10769-486 10769-388 10769-408 10769-406 10018-928 10769-396 10769-474 10769-452 10769-394 10769-472 10769-450 10769-392 10769-470 10769-492 10769-390 10769-490 10769-438 10769-416 10769-458 10769-436 10769-414 10769-478 10769-434 10769-412 10769-456 10769-398 10769-410 10769-454 10769-476 10769-432 10769-440 10769-462 10769-484 10769-482 10769-480
SpillStopper Work Surfaces, Labconco
Vitrinas de gases y recintos Campanas de gases químicos
SpillStopper Work Surfaces are molded from a special formulation of corrosion-resistant epoxy resins
Dished and contoured to conform to the interior liner of Protector Hoods. Front edge has a large radius to aerodynamically direct airflow into the hood. Pre-drilled 1.5" (5.8 cm) diameter holes for venting and 2.5" (6.4 cm) diameter holes for plumbing pass-through.
Información para pedidos: May be ordered with a pre-cut 15.2 x 7.6 cm (6 x 3") oval cupsink cutout. Cupsink is sold separately. SpillStopper Work Surfaces provide support for Protector Premier, XStream, XL, Echo and Airo Laboratory Hoods.