VWR® Media/Storage Bottles with GL Screw Caps
Proveedor: VWR
VWR®
1632414631956 1632414631950 1632414632100 1632414631050 1632414632250 1632414631100 1632414632956 1632414632950 1632414631940 1632414540956 1632414631968 1632414632500 1632414632966 1632414631954 1632414631500 1632414632940 1632414631966 1632414540966 1632414631250
10754-818CS 1258.44 MXN
76316-244 10754-818 10754-828 75993-584 75993-580 10754-834 10754-822 75993-582 10754-832 10754-820 10754-838 10754-816 10754-826 10754-814 10754-836 10754-824 10754-830 10754-840 76316-246
VWR® Media/Storage Bottles with GL Screw Caps
Frascos Botellas de vidrio
Media/storage bottles are designed with a wide opening (30 mm Int.Ø) and a drip-free polypropylene pour ring.
  • Very good chemical resistance
  • High thermal-shock resistance
  • Scale graduations
  • Autoclavable
It can be autoclaved to 180 °C (356 °F). They feature enhanced graduations and marking spots printed in permanent white color.
Certificaciones: These bottles are manufactured from high quality Type 1, Class A, 33 expansion Borosilicare glass.
Información para pedidos: All bottles are available with or without a GL 45 polypropylene cap, with the exception of the 50 ml bottles, which feature a GL 32 thread.