VWR® Snap-Cap Centrifuge Tube, 5 ml
Proveedor: VWR
VWR®
C1005-T5-ST C1005-T5AST C1005-T5
10002-728PK 971.45 MXN
10002-731 10002-740 10002-728
VWR® Snap-Cap Centrifuge Tube, 5 ml
Tubos Tubos de centrífuga
VWR® Snap-Cap Centrifuge tubes are made of high-clarity, low-binding, virgin polypropylene.
  • Constructed of high clarity, low-binding, virgin polypropylene
  • Tubes can withstand up to 25000 ×g
  • Supplied with an attached cap with a tight, leak-proof seal
  • Certified RNase- and DNase-free
The tubes have a 16 mm Ø, which is identical to standard 15 ml tubes. This ensures compatibility with existing centrifuge rotors.
Tubes feature dual gradations at every 0.2 and 0.5 ml level. They are supplied with an attached cap that provides a tight, leak-proof seal. Caps can be easily opened with one hand, and provide a secure seal from –86 to 80 °C (–122 to 176 °F). The center of the cap has a thin membrane area for easy access by a syringe or needle.
Información para pedidos: The assorted color collection of VWR® Snap-Cap Centrifuge Tubes includes 50 each of red, blue, green, and yellow tubes.