Order Entry
Cannon-Fenske Routine Viscometer, Cannon Instrument Company
undefined
Cannon-Fenske Routine Viscometer, Cannon Instrument Company
  •  
    Size
    Range
    Approx. constant
    Supplier No.
  • Calibrated
    Calibrated
    47734-928
    9721-B50
    25
    0.5 - 2 cSt
    0.002 cSt/s
    9721-B50
    Each (1UN)
    Each (1UN) - $6,086.41
    $6,086.41
    Each (1UN)
       $6,086.41
    Small Business Enterprise
    Specifications
    Environmentally Preferable
    Small Business Enterprise
  • Calibrated
    Calibrated
    47734-936
    9721-B62
    150
    7 - 35 cSt
    0.035 cSt/s
    9721-B62
    Each (1UN)
    Each (1UN) - $5,709.05
    $5,709.05
    Each (1UN)
       $5,709.05
    Small Business Enterprise
    Specifications
    Environmentally Preferable
    Small Business Enterprise
  • Calibrated
    Calibrated
    47734-940
    9721-B68
    300
    50 - 250 cSt
    0.25 cSt/s
    9721-B68
    Each (1UN)
    Each (1UN) - $5,709.05
    $5,709.05
    Each (1UN)
       $5,709.05
    Small Business Enterprise
    Specifications
    Environmentally Preferable
    Small Business Enterprise
  • Calibrated
    Calibrated
    47734-944
    9721-B74
    400
    240 - 1200 cSt
    1.2 cSt/s
    9721-B74
    Each (1UN)
    Each (1UN) - $5,709.05
    $5,709.05
    Each (1UN)
       $5,709.05
    Small Business Enterprise
    Specifications
    Environmentally Preferable
    Small Business Enterprise