VWR® Round Carboy, Narrow Mouth
Proveedor: VWR
VWR®
583371 583377 583372 583322 583321 583260 583270 583376 583373 583250
10755-086CS 10791.12 MXN
10755-086 10755-110 10755-098 75795-688 10755-112 75795-674 10755-114 10755-124 10755-126 10755-084
VWR® Round Carboy, Narrow Mouth
Frascos Garrafones
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene, LDPE, and HDPE formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. LDPE and HDPE carboys are not autoclavable.
Certificaciones: Resin meets US FDA 21 CFR, USP Class VI.