VWR® Round Carboy with Spigot, Narrow Mouth
Proveedor: VWR
VWR®
583290 583300 584401 584403 584380 584304 584400 584390 583280
10755-130EA 6592.37 MXN
10755-130 75795-676 75795-698 10755-088 75795-682 10755-116 10755-118 10755-128 10755-090
VWR® Round Carboy with Spigot, Narrow Mouth
Frascos Garrafones
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non sterile
  • Polypropylene, LDPE, and HDPE formats available
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof on standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. LDPE and HDPE carboys are not autoclavable.
Certificaciones: Resin meets US FDA 21 CFR, USP Class VI.
Información para pedidos: Spigot included. Always disassemble spigot and cap from carboy before autoclaving.