SteriPlast® Sampling Scoop, Bürkle
Proveedor: Bürkle
SteriPlast® SteriPlast
5378-1013 5378-1020 5378-1007 5378-1004 5378-1005 5378-1002 5378-1016 5378-5018 5378-5014 5378-1006 5378-1019 5378-1003 5378-1015 5378-1008 5378-1014 5378-1001
10769-658PK 931.53 MXN
10769-658 10810-867 10769-666 10769-656 10810-869 10769-664 10769-654 10769-662 10770-178 75848-662 10810-761 10769-660 76501-360 10810-763 76501-392 76501-370
SteriPlast® Sampling Scoop, Bürkle
Palas
These scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed, gamma radiation sterilised
  • Production, assembling and packaging acc. to ISO cleanroom class 7
They are also available with a transparent lid to close it immediately after the sampling procedure to secure a sterile and contamination-free sample. Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.
Certificaciones: Complies with EU food requirements and FDA regulations.