Laboratory Line Vertical Autoclaves, Heidolph Tuttnauer
Proveedor: Heidolph
023210745CA 023210745 023210744 023210990
89423-406EA 1242226.18 MXN
10001-546 CA11028-312 89423-406 10048-728
Laboratory Line Vertical Autoclaves, Heidolph Tuttnauer
Esterilizadores Autoclaves (Esterilizadores de vapor)
These products cover a wide range of applications for laboratories in research institutes, universities, pharmaceutical, food, chemical, and biotechnological industries.
  • Maximum temperature of 135 °C (275 °F)
The units feature an advanced microprocessor control panel and chamber made of 316L or 316Ti stainless steel. For life science applications, the Heidolph Tuttnauer line successfully meets the challenges in modern laboratories with a flexible range of features and a sophisticated control system.
Units includes a PID (proportional integral differential) pressure control feature and the ability to store the last 200 cycles in built-in memory. Up to 6 temperature sensors and 4 pressure sensors can be connected. Users can have up to 30 identification codes and passwords for access level control. The controller and software comply with the 21 CFR part 11 standard. Units have a sterilization temperature range 1050 to 1370 °C (1922 to 2498 °F).
These mobile units feature plug and play operation only with an electric power connection. There is no need to connect autoclave to building utilities for distilled water, tap water, and drain. Supplied with wheels that lock into place.
The advanced Laboratory Line provides a single solution for the full spectrum of sterilization needs including liquids, culture media, instruments, glassware, plastics, pipette tips, biological waste, contaminated media, and other laboratory items.
Información para pedidos: This unit include a set of 2 sample temperature probes, internal thermometer, water-sensing electrode, external pressure gauge, printer, and USB connection.