Aluminium Dishes, with Flat bottom and Straight Sides, Thermo Scientific
Proveedor: Thermo Scientific
39069-UD 39073-KH 39069-KH 39077-UD 39077-KH 39083-KH 39074-UD 39084-KH 39079-KH 39074-KH 39073-UD 39079-UD 039072-UD 39084-UD 39076-KH 39080-KH 39080-UD 039072-KH
AA39080-UDEA 1739.9 MXN
AA39080-UD AA39073-KH AA39072-KH AA39076-KH AA39084-UD AA39069-UD AA39077-KH AA39074-KH AA39079-KH AA39080-KH AA39074-UD AA39073-UD AA39072-UD AA39069-KH AA39077-UD AA39083-KH AA39084-KH AA39079-UD
Aluminium Dishes, with Flat bottom and Straight Sides, Thermo Scientific
Bandejas/Platos/Ollas Cápsulas de evaporación
These beakers are made up of aluminium
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Designed to provide long life, increase efficiency and lower the running cost.