VWR® Lead-Free Autoclave Tapes
Proveedor: VWR
VWR®
VWR-T251-47 VWR-T254-47 VWR-T151-47 VWR-T351-47 VWR-T156-47 VWR-T353-47
10127-458CS 1007.76 MXN
10127-458 10127-460 76376-470 10127-462 10127-464 76376-496
VWR® Lead-Free Autoclave Tapes
Indicadores de esterilización
VWR® Lead-free autoclave tape is sterilization package sealing tape with a process indicator.
  • Lead-free
  • Autoclavable
  • Sticks securely under high-temperature and high-pressure conditions
Stripes turn from light green to black after exposure to steam sterilization conditions, and the tape sticks securely under high-temperature and high-pressure conditions. Tape will remove cleanly and easily following sterilization.
Designed for gravity sterilizers operating at 121 °C (250 °F), pre-vacuum steam sterilizers operating at 134 °C (273 °F), and formaldehyde/alcohol chemical sterilizers.
This tape can be used in temperatures ranging from -23 to 121°C (-9 to 250°F). Designed for gravity sterilizers and pre-vacuum steam sterilizers.