Once-A-Day® Vertos™ Bowie-Dick Test, Propper Manufacturing
Proveedor: Propper
Once-A-Day®
26610100
10798-528CS 5654.33 MXN
10798-528
Once-A-Day® Vertos™ Bowie-Dick Test, Propper Manufacturing
Indicadores de esterilización
Once-A-Day® Vertos™ bowie dick test is an eco-friendly, lead-free option for daily testing.
  • Disposable bowie-dick test pack
  • Lead and heavy metal free ink
  • Distinct blue to pink color change
This test provides a challenge for pre-vacuum sterilizers at the start of each day, or after routine or other maintenance. Helps in detecting at low temperature/pressure conditions, wet and superheated steam. For use in pre-vacuum steam sterilizers at 270 to 276 °F.
Certificaciones: AAMI / ISO 11140-1:2010, Type 2 AAMI / ISO 11140-5:2007