General Purpose Viscosity Standards, Silicone Fluids, BrookField Engineering
Proveedor: Brookfield
60000 CPS
100241-370EA 3290.8 MXN
100241-370
General Purpose Viscosity Standards, Silicone Fluids, BrookField Engineering
Normativas Estándar para viscometría
These viscosity standards is used to verify calibration of laboratory viscometers/rheometers.
  • Accurate to ±1% of viscosity value
  • Economical
  • Excellent temperature stability
The selection of one or two fluids will normally provide sufficient measurement points to verify calibration of your instrument.
1 cP=1 mPa s
Certificaciones: NIST traceable certification