VWR® TraceClean® Wide Mouth Glass Packer Bottles, Amber
Proveedor: VWR
QLAP15900279 QLAP15900275 QLAP15900277 QLAP15900281 QLAP15900257
89126-600CS 2350.43 MXN
89126-600 89126-602 89126-576 89126-604 89126-606
VWR® TraceClean® Wide Mouth Glass Packer Bottles, Amber
Frascos Botellas de vidrio
These TraceClean® glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing semi-volatiles (light sensitive compounds), pesticides, PCBs, diesel range organics, TPH, Metals, and phenolics.
  • Amber glass fabrication
  • Recommended for UV light-sensitive samples
  • Wide-mouthed design
Secondary uses include containment, bench vessels, and archiving.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
Certificaciones: QAL.
Información para pedidos: Container measurements and capacity 'contain' volumes may vary.