Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Proveedor: Ohaus
80780029 80780021 80780022 80780028 80780023 80780020 80780024 80780027
11384-132EA 3803.28 MXN
11384-120 11384-130 11384-122 11384-132 11384-124 11384-134 11384-126 11384-136
Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Pesas Pesas de calibración
Manufactured to international standards, individual calibration weights are available for a variety of precision applications
Calibrated to ASTM Class 1. Extra-fine accuracy for calibrating high precision analytical balances with a readability as low as 0.1 to 0.01mg.