Dialysis Tubing
Proveedor: ALDON CORP SE
ADDD0106-100 ADDD0106-10 ADDD0108-50 ADDD0105-25 ADDD0107-10 ADDD0106500FT ADDD0107-100 ADDD0105-10 ADDD0109-100 ADDD0108-10 ADDD0109-10 ADDD0105-100 ADDD0108-100
470163-408EA 407.58 MXN
470163-408 470163-410 470163-402 470163-424 470163-414 470231-460 470163-400 470163-412 470163-406 470163-418 470163-404 470163-426 470163-416
Dialysis Tubing
Tuberías Tuberías para diálisis
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
  • Available in multiple lengths