ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Proveedor: ERGOMAT INC
ERGOMAT®
EH0408-BLU EH0218-BLU EH0407-BLU EH0210-BLU EH0204-BLU EH0316-BLU EH0313-BLU EH0206-BLU EH0418-BLU EH0404-BLU EH0407-BLU EH0415-BLU EH0203-BLU EH0214-BLU EH0219-BLU EH0420-BLU EH0311-BLU EH0212-BLU EH0305-BLU EH0212-BLU EH0416-BLU EH0218-BLU EH0316-BLU EH0206-BLU EH0405-BLU EH0411-BLU EH0216-BLU EH0419-BLU EH0406-BLU EH0217-BLU EH0210-BLU EH0213-BLU EH0411-BLU EH0305-BLU EH0211-BLU EH0420-BLU EH0215-BLU EH0319-BLU EH0209-BLU EH0318-BLU EH0306-BLU EH0306-BLU EH0217-BLU EH0308-BLU EH0312-BLU EH0310-BLU EH0405-BLU EH0205-BLU EH0418-BLU EH0317-BLU EH0220-BLU EH0408-BLU EH0410-BLU EH0303-BLU EH0312-BLU EH0208-BLU EH0414-BLU EH0415-BLU EH0309-BLU EH0417-BLU EH0309-BLU EH0303-BLU EH0413-BLU EH0412-BLU EH0416-BLU EH0313-BLU EH0320-BLU EH0318-BLU EH0205-BLU EH0319-BLU EH0320-BLU EH0207-BLU EH0417-BLU EH0208-BLU EH0307-BLU EH0304-BLU EH0314-BLU EH0315-BLU EH0204-BLU EH0409-BLU EH0207-BLU EH0308-BLU EH0413-BLU EH0216-BLU EH0203-BLU EH0214-BLU EH0410-BLU EH0213-BLU EH0414-BLU EH0307-BLU EH0406-BLU EH0317-BLU EH0304-BLU EH0209-BLU EH0219-BLU EH0220-BLU EH0314-BLU EH0409-BLU EH0211-BLU EH0404-BLU EH0419-BLU EH0311-BLU EH0215-BLU EH0310-BLU EH0412-BLU EH0315-BLU
22231-150EA 4982.9 MXN
CA89169-658 89169-648 22231-154 22231-152 22231-150 89169-654 CA89169-662 89169-652 CA89169-660 CA89169-700 CA89169-666 89169-658 CA89169-664 89169-656 89169-650 CA89169-704 CA89169-702 CA89169-668 CA89169-706 89169-664 CA89169-674 CA89169-672 89169-662 89169-668 CA89169-678 89169-702 CA89169-676 89169-666 89169-700 CA89169-670 89169-660 89169-706 89169-704 89169-676 CA89169-684 89169-674 CA89169-682 CA89169-688 89169-678 CA89169-686 CA89169-680 89169-672 89169-670 CA89169-608 89169-686 CA89169-696 CA89169-694 89169-684 CA89169-612 CA89169-698 89169-688 CA89169-610 89169-682 CA89169-692 89169-680 CA89169-690 CA89169-616 CA89169-614 89169-608 CA89169-618 89169-610 89169-698 89169-696 CA89169-622 89169-614 89169-612 CA89169-620 89169-690 89169-694 89169-692 89169-618 CA89169-626 89169-616 CA89169-624 CA89169-628 89169-620 CA89169-630 CA89169-634 89169-624 89169-622 CA89169-632 89169-628 CA89169-638 89169-626 CA89169-636 89169-632 CA89169-640 89169-630 89169-636 CA89169-644 89169-634 CA89169-642 CA89169-648 CA22231-154 CA89169-646 89169-638 CA22231-152 CA22231-150 89169-642 CA89169-652 89169-640 CA89169-650 CA89169-656 89169-646 CA89169-654 89169-644
ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Alfombrillas Alfombrillas antifatiga
The ERGOMAT® Hygiene is a cone profile ergonomic mat that is well suited for use in critical environments requiring daily caustic sanitation.
  • Hygiene mats have a broad range of pH resistance (2 to 13), and tolerate a variety of chemical exposures (except oils)
  • Mats can be used in hospitals and clinics, laboratories, food processing facilities, and commercial kitchens
  • Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
  • Mats can be safely autoclaved (2’×3’ size only)
Mats are 11 mm (7/16") thick and 91.4 cm (36") wide, with beveled edges on all sides to prevent curling and eliminate trip hazards. These nonslip rubber mats promote occupational safety and health and deliver ergonomic benefits in the most demanding environments.
Certificaciones: They are USDA approved, meet CIFA standards for food processing and are ISO Class 5 cleanroom rated.
Información para pedidos: Mats come with manufacturer’s three-year warranty.