4' Purifier® Logic®+ Class II A2 Biosafety Cabinets, International Electrical Configurations, Labconco®
302481100 302480160 302481060 302480060 302481160
85002-568EA 358358 MXN
89412-912 89412-906 89412-908 89412-910 85002-568
4' Purifier® Logic®+ Class II A2 Biosafety Cabinets, International Electrical Configurations, Labconco®
The new Purifier Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality.
  • Intuitive programming
  • Color-coded menus
  • Status messages and alarms
  • Diagnostic troubleshooting
  • Provides personnel, product and environmental protection
  • Suitable for work in biosafety levels 1, 2 and 3
Advanced ergonomic design has shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system
Using the same ECM blower technology as the Purifier Logic®, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. These biohazard cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. In addition, some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. Please refer to sash opening, electrical configuration and product specifications for details.
MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.