Aluminum Single Element ICP and ICP/MS Certified Reference Standards, BDH® ARISTAR®
BDHCGALCL1-1 BDHCGAL1-1 89800-002 89800-264 89800-004 BDHMSAL-10 BDHMSAL-100 BDHMSAL-10 BDHCGAL1-1 BDHCGALCL1-1 89800-266 BDHMSAL-100
82025-940EA 1013.64 MXN
89800-264 82026-122 82025-940 CA82026-120 CA82025-940 82026-120 82025-942 89800-004 CA82025-942 89800-002 89800-266 CA82026-122
Aluminum Single Element ICP and ICP/MS Certified Reference Standards, BDH® ARISTAR®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125mL and 500mL sizes packaged in plastic LDPE bottles.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.