Single Element ICP and ICP/MS Certified Reference Standards, Calcium, ARISTAR®, VWR Chemicals BDH®
BDHCGCA1-1 BDHMSCA-10 89800-038 89800-298 BDHCGCA1-1 BDHMSCA-100 89800-296 BDHMSCA-10 BDHMSCA-100
82025-960EA 1428.61 MXN
CA82025-960 82026-144 89800-298 89800-296 89800-038 CA82026-146 82026-146 82025-960 CA82026-144
Single Element ICP and ICP/MS Certified Reference Standards, Calcium, ARISTAR®, VWR Chemicals BDH®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Concentrations ranging from 10, 100, 1,000, and 10,000 ppm
  • Engineered to be stable, compatible, traceable to NIST SRMs
  • Manufactured and tested under ISO Guide 34 & ISO 17025 guidelines
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.
Información de suministro: Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online.