Single Element ICP and ICP/MS Certified Reference Standards, Chromium, ARISTAR®, VWR Chemicals BDH®
BDHMSCR3-100 89800-046 BDHMSCR3-100 89800-216 BDHCGCR61-1 89800-048 89800-312 89800-244 BDHCGCR31-1 BDHCGCR31-1 BDHMSCR3-10 BDHCGCR61-1 BDHMSCR3-10 89800-314
82025-968EA 1154.42 MXN
82026-156 89800-048 89800-312 89800-046 89800-244 82025-970 CA82025-970 82025-968 CA82025-968 89800-314 89800-216 CA82026-158 CA82026-156 82026-158
Single Element ICP and ICP/MS Certified Reference Standards, Chromium, ARISTAR®, VWR Chemicals BDH®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125 mL and 500 mL sizes packaged in plastic LDPE bottles.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.
Precaución: 1000 ppm = 1000 µg/ml = 1000 mg/l