Single Element ICP and ICP/MS Certified Reference Standards, Copper, ARISTAR®, VWR Chemicals BDH®
BDHCGCU1-1 BDHMSCU-10 89800-052 89800-322 BDHMSCU-100 BDHMSCU-10 BDHMSCU-100 89800-320 BDHCGCU1-1
82025-974EA 1428.61 MXN
89800-320 82025-974 CA82026-162 82026-162 CA82025-974 CA82026-160 82026-160 89800-322 89800-052
Single Element ICP and ICP/MS Certified Reference Standards, Copper, ARISTAR®, VWR Chemicals BDH®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125mL and 500mL sizes packaged in plastic LDPE bottles.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.