Single Element ICP and ICP/MS Certified Reference Standards, Silicon, ARISTAR®, VWR Chemicals BDH®
89800-230 89800-156 89800-258 BDHCGSI1-1 89800-158 89800-154 89800-464 89800-466 BDHCGSI1-1
82026-058EA 1013.64 MXN
89800-154 89800-230 89800-158 89800-466 89800-258 89800-156 89800-464 CA82026-058 82026-058
Single Element ICP and ICP/MS Certified Reference Standards, Silicon, ARISTAR®, VWR Chemicals BDH®
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125mL and 500mL sizes packaged in plastic LDPE bottles.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 ppm.