BootieButler® Shoe Covers, Protexer
Proveedor: Protexer
BootieButler®
KBNS40-600-ESD KBNS40-600-FAS KBNS40-600 KBNS28-750 KBNS28-750-FAS KBNS28-1-ESD KBNS28-1-FAS KBNS28-750-ESD
10836-051CS 9742.19 MXN
75796-908 75798-788 75798-786 10836-051 75798-784 76186-138 75798-782 75798-790
BootieButler® Shoe Covers, Protexer
Prendas desechables
The original BootieButler® Shoe Cover is an economical anti-skid and anti-static shoe cover that serves well in a variety of situations
  • Super anti-skid, slip-resistant pattern
  • Antistatic material
  • Lightweight and durable for extra-long wear
  • One size fits most
  • Specifically designed for use in the BootieButler® automatic dispenser
BootieButler® Shoe Covers are ideal for use in cleanrooms, food processing, real estate, healthcare, construction, manufacturing, research and development, and energy applications.
Constructed of durable spunbond polypropylene material with non-skid, non-conductive soles. The skid-resistant pattern improves friction and traction. A strong elastic band around the ankle allows for a universal fit.
Certificaciones: Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5.
Información para pedidos: These shoe covers are designed to be used in the BootieButler® automatic dispenser, and not as a bulk product.