VWR® Series Glass Door Chromatography Refrigerators with Natural Refrigerants
HCCS-35
10819-306
VWR® Series Glass Door Chromatography Refrigerators with Natural Refrigerants
Units feature a digital microprocessor temperature controller, allowing for the precise temperature management necessary for critical samples and supplies.
  • Microprocessor temperature controller
  • Sample simulated digital temperature display
  • Audible and visual hgh/low temperature alarms
  • Interior duplex electrical outlet
  • Two, 2” gasketed side access ports
  • Keyed door locks
  • Swivel casters adds 4” to height
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
An automatic cycle-defrost design prevents product damaging temperature spikes.
Certificaciones: UL/C-UL listed.
Información para pedidos: Two-year parts and labor warranty, five years on compressor parts.