VWR® Performance Series Chromatography Refrigerators with Natural Refrigerants
VWR®
HCCP-49-TS
10819-616EA 289361 MXN
10819-616
VWR® Performance Series Chromatography Refrigerators with Natural Refrigerants
Refrigeradores
The VWR® Performance Series Chromatography Refrigerator units are purpose built for scientific and medical applications requiring superior temperature management and state of the art product security and data logging.
  • Monitoring and alarms
  • Sample-simulated temperature display
  • Full alarm package
  • Alarm mute with ring back
  • Adjustable volume
  • Alarm battery backup
  • Remote contacts
  • Alarm validation
  • Swivel casters (adds four inches to height)
The all-new Touch Screen temperature display and alarm module offers the latest in advanced temperature control and recording of temperature history.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Certificaciones: UL/C-UL listed.
Información para pedidos: Three-year parts and labor warranty, five years on compressor parts.