VWR® Plus Series Solid Door Laboratory Refrigerators
SHCLS-72 SHCLS-16 SHCLS-23 SHCLS-49 SHCLS-26 SHCLS-36 SHCLS-12
76169-592 76169-618 76169-580 76169-596 76169-594 76169-612 76169-610
VWR® Plus Series Solid Door Laboratory Refrigerators
The VWR® Plus Series Solid Door Laboratory Refrigerators are purpose built for scientific and medical applications.
  • Temperature controller
  • Digital setting accuracy
  • Precise temperature maintenance
  • Independent operation for security
  • Monitoring and alarms
  • Sample-simulated temperature display
  • High/low temperature alarm
  • Sensor error alarm
  • Power failure alarm
  • Alarm mute with ring back
  • Remote contacts
  • Alarm validation
  • Resettable 1-point min/max history
Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes.
Certificaciones: UL/C-UL listed (except for 76169-610, 76169-592, and 76169-618, which are ETL listed).
Información para pedidos: Unit comes with casters with footed brake. Two-year parts and labor warranty, five years on compressor parts.
Precaución: (Note: swivel casters add 3½" to height, which is not included in dimensions)