VWR® Plus Series Solid/Glass Door Compact Laboratory Refrigerators with Natural Refrigerants
Proveedor: VWR
VWR®
HCLP-10G
10819-782EA 4617.67 MXN
10819-782
VWR® Plus Series Solid/Glass Door Compact Laboratory Refrigerators with Natural Refrigerants
Refrigeradores
The VWR® Plus Series Solid/Glass Door Compact Laboratory Refirgerators feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
  • Temperature Controller
  • Digital setting accuracy
  • Precise temperature maintenance
  • Monitoring and alarms
  • Digital temperature display
  • High/low temperature alarms
  • Remote alarm contacts
Uniformed forced air directional cooling
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
The all new digital temperature display and alarm module offers industry leading product security with a monitoring and alarm system that is 100% independent of the temperature controller. Real time sample simulated temperatures are constantly displayed and supported by audible/visual high and low temperature alarms, along with power outage and sensor error alarms. Standard features include: Keyed door locks, probe access port.
Certificaciones: UL/C-UL listed.
Información para pedidos: Two-year parts and labor warranty, five years on compressor parts.