VWR® Plus Series Manual Defrost Laboratory Freezers
Proveedor: VWR
VWR®
MFS-30 MFS-3030
10820-410 10820-406
VWR® Plus Series Manual Defrost Laboratory Freezers
Congeladores Congeladores de laboratorio/aplicaciónes generales
The VWR® Plus Series Manual Defrost Laboratory Freezer units are purpose built for critical cold storage applications, but where built-in data logging functionality may not be a necessity.
  • Monitoring and alarms
  • Sample-simulated temperature display
  • High/low temperature alarm
  • Sensor error alarm
  • Power failure alarm
  • Alarm mute with ring back
  • Remote alarm contacts
  • Alarm validation
  • Resettable 1-point min/max history
  • Temperature controller
  • Digital setting accuracy
  • Precise temperature maintenance
  • Independent operation for security
  • Caster platform adds four inches to height
  • Inner "fast-freeze compartments" with doors for temperature maintenance and recovery
The all-new temperature display and alarm module offers an array of robust security features with a monitoring and alarm.
Certificaciones: UL/C-UL listed.
Información para pedidos: Two-year parts and labor warranty, five years on compressor parts.
Precaución: Defrosting of the unit will need to be performed periodically to prevent severe ice build-up.