VWR® Standard Manual Defrost Laboratory Series Freezers
Proveedor: VWR
VWR®
MFV-30
10819-410EA 125626 MXN
10819-410
VWR® Standard Manual Defrost Laboratory Series Freezers
Congeladores Congeladores de laboratorio/aplicaciónes generales
The VWR® Standard Series Manual Defrost Laboratory Series Freezers are designed specifically with general-purpose applications in mind.
  • Adjustable operating temperature range: –15 to –25 °C (5 to –13 °F)
  • Direct-cool fixed evaporator shelves
These units are ideal when stable temperatures averaging -20°C are required but precise temperature control, maintenance, and alarms are not necessary. These freezers feature a manual defrost design to eliminate damaging temperature spikes from defrost cycles. This ensures valuable sample security. Defrosting of the unit must be performed periodically to prevent severe ice build-up.
Freezer 10819-410 comes with leveling legs (not casters). An optional roller base platform is available (97043-518).
Certificaciones: UL/C-UL listed.
Información para pedidos: Two-year parts and labor warranty, five years on compressor parts.