Deep Well Plates, Axygen® Scientific
Proveedor: Axygen
P-96-450V-C P-96-450R-C P-96-450V-C-S P-96-450R-C
47743-982CS 2139.26 MXN
490005-358 47743-996 47743-998 47743-982
Deep Well Plates, Axygen® Scientific
Microplacas Microplacas HTS
Axygen® Deep Well and Assay plates are used for sample collection, in vitro growth chambers, and long-term storage.
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to –80 °C.