PVAmicro Cloth, Vileda Professional-FHP
Proveedor: Vileda Professional
143587 143588 143585 143586
10153-212CS 204.96 MXN
10153-212 10153-210 10153-220 10153-218
PVAmicro Cloth, Vileda Professional-FHP
Toallitas Paños de limpieza reutilizables Paños para aplicaciones generales
PVAmicro is an innovative all-purpose cloth made from microfiber fabric impregnated with PVA.
  • PVAmicro holds 40 times less particle residue after rinsing, compared to ordinary knitted microfibre cloth
  • Cloth can be washed repeatedly
  • Ideal for cleaning all types of hard surfaces, such as office furniture, glass, or stainless steel
The PVA impregnation makes the cloth very easy to rinse and wring. PVAmicro achieves a perfect, shiny cleaning result without streaks every time.
Información para pedidos: PVAmicro cloth is packaged 5 cloths per pack, 20 packs per case.