VWR® Plus Laboratory Refrigerator Freezer Combination Units
Proveedor: Avantor
HCRFC7
10819-928
VWR® Plus Laboratory Refrigerator Freezer Combination Units
The VWR® Plus Series Laboratory Refrigerator/Freezer Combination units offer a variety of sizes and configurations to provide a solution where space is at a premium.
  • Direct-cool fixed evaporator shelves in freezer
  • Forced air directional cooling in refrigerator
  • Environmentally friendly refrigerants
Units marked with manual defrost freezer compartments will need to be defrosted periodically to prevent severe ice build-up. Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines. All units have temperature range: fridge: 1 to 10 ºC and freezer: -15 to -25 ºC.
Certificaciones: UL/C-UL listed.
Información para pedidos: Two-year parts and labor warranty, five years on compressor parts. (Please note the 1 year extended warranty 10753-492 needs to be purchased for the freezer and the refrigerator separately, so 2x 10753-492 need to be ordered to cover both parts.)
Precaución: Units marked as “freestanding” are not designed for built-in and/or enclosed installations. Improper installation will void the warranty and will lead to temperature maintenance issues, overheating and possible failure.