VWR® Standard Series Hazardous Location Explosion Proof Freezers
VWR®
EFV-30
10819-916
VWR® Standard Series Hazardous Location Explosion Proof Freezers
Congeladores Congeladores de material inflamable
The VWR® Standard Series Explosion Proof Freezers are designed to meet NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D.
  • All electrical components are housed externally
  • For use in volatile or explosive conditions
  • Not intended for common labs
These units are engineered for use in environments where volatile or explosive conditions could potentially exist. They are not intended for common lab / room environments that are not designed to be “explosion-proof” (See: Flammable Refrigerators and Freezers). All electrical components are housed outside of the unit and sealed per NFPA and OSHA guidelines.
Certificaciones: UL/C-UL listed.
Información para pedidos: Not equipped with line cord and plug; hardwired into appropriate conduit box. two-year parts and labor warranty.
Precaución: Warning: Units must be hard-wired in conduit to 120VAC power source. Power cords with plugs are not acceptable.