Precision™ Coliform Water Baths, Thermo Scientific
Proveedor: Thermo Scientific
TSCOL35 TSCOL19
10805-318EA 65623.03 MXN
10805-318 10805-316
Precision™ Coliform Water Baths, Thermo Scientific
Baños Baños para coliformes
Support your fecal coliform testing with Thermo Scientific™ Precision™ coliform water baths, designed specifically for this application
Baths are preset to optimal temperatures of 35.0, 41.5, 44.5 and 45.5°C for easy use. Each bath is factory is factory calibrated, with later field calibration enabled from the front panel without the need for tools. Benefit from outstanding chemical and corrosion resistance with epoxy powder-coated exterior, and easily clean the chamber with its seamless stainless-steel interior.
Certificaciones: cULus Listed, CE compliant, US FDA Class I Medical Device.
Información para pedidos: Included Stainless Steel Lid, Diffuser Tray, Power Cord, Yellow Duck.