PYREX® Volumetric Pipettes, Class A, Color-Coded with Colored Graduations, Corning
Proveedor: Corning
PYREX®
7103-15 7103-7 7103-4 7103-25 7103-10 7103-1 7103-2 7103-6 7103-8 7103-3 7103-20
33500-538CS 1600 MXN
33500-552 33500-542 33500-550 33500-540 33500-556 33500-546 33500-544 33500-538 33500-558 33500-548 33500-560
PYREX® Volumetric Pipettes, Class A, Color-Coded with Colored Graduations, Corning
Pipetas Pipetas graduadas
These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.
  • Reusable
  • Color-coded with colored graduations and calibration lines enameled onto the glass