RPMI-1640 Mammalian Cell Culture Medium, HiMedia
Proveedor: HiMedia Laboratories
AT028-1L AT028-1L AT060-1L AT060-1L AT112A-10L AT112A-10L AT112A-20L AL028G-2X500ML AT028-5L AT112A-20L AL060G-2X500ML AT028-5L AT028-20L AL060G-5X500ML AT060-5L AL028G-5X500ML AT060-5L AT028-20L
95062-768EA 1719.99 MXN
95016-630 95016-632 95057-922 CA95016-632 95062-770 CA95016-630 CA95016-624 95016-624 CA95057-924 CA95057-922 95062-762 95057-924 95016-626 CA95016-628 95016-628 95062-764 CA95016-626 95062-768
RPMI-1640 Mammalian Cell Culture Medium, HiMedia
Medios de cultivo celular
Roswell Park Memorial Institute (RPMI) media are a series of media developed by Moore et al for the culture of human normal and neoplastic cells in vitro.
Extensively used for a wide range of anchorage dependant cell lines
Needs to be supplemented with 5-20% fetal bovine serum
Also known to support growth of cells in the absence of serum
RPMI 1640 is the most commonly used medium in the series. A modification of McCoy’s 5A medium, the medium was specifically designed to support the growth of human lymphoblastoid cells in suspension culture. Presently the medium is extensively used for a wide range of anchorage dependant cell lines. The medium needs to be supplemented with 5-20% fetal bovine serum. The medium is also known to support growth of cells in the absence of serum.