PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Proveedor: Corning
PYREX®
4000-10024 4000-25024 4000-10019
26307-264 26307-262 26307-270
PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Trampas de vacío
These PYREX® traps are used in rotary evaporators. They have a round body with upper and lower stems.
  • Lower stem has a Standard Taper inner joint at the bottom and extends upward inside the body
  • The closed top of this stem has two side openings
  • The upper stem has a Standard Taper outer joint