Protector® Base Cabinet Filler Panels, Labconco®
Proveedor: Labcono
Protector®
992031010814 992030610814
89238-198EA 3457.07 MXN
89238-198 89238-200
Protector® Base Cabinet Filler Panels, Labconco®
Mobiliario Mobiliario de laboratorio
One 8" filler panel is included with each storage cabinet to extend cabinet depth to 30" and to enclose the utility chase behind the cabinet.
  • Available in 8", 14", 15.5", and 20" depths
  • Available for standard height and ADA height cabinets
Cabinets supporting hoods with external depths greater than 31.7" require deeper filler panels.
For use with Labconco® Protector® Acid Storage Cabinets, Protector® Solvent Cabinets, Protector® Standard Cabinets, and Protector® Vacuum Pump Storage Cabinets.