VWR® CE Flat Sponge Mop Heads
Proveedor: VWR
150264 150266 148962
89032-166CS 17504.95 MXN
89032-166 10208-978 89032-170
VWR® CE Flat Sponge Mop Heads
Mopas
VWR® CE Flat Sponge Mop Heads are manufactured and sterlized in the USA.
  • Polyester lamination allows for tear-free application of disinfectants, cleaning, and spill absorption
  • Polyester allows for optimized manufacturer recommended contact times for disinfectants
  • Unlike competitive products, this polyester is laminated to the foam without the use of adhesives, which could contaminate surfaces
  • Fits common plastic and stainless steel flat mopping frames
  • Autoclavable
Sterile heads are validated to 10-6 SAL per ISO 11137. Mop heads are individually double bagged, with Lot number and sterile indicator dot on each package (where applicable). A certificate of sterility is available upon request with each shipment.