Septa Cap for Vials, C&G Containers
Proveedor: C&G Containers
CAH242005I04
71000-006EA 11.71 MXN
71000-006
Septa Cap for Vials, C&G Containers
Tapas Tapones para viales de cromatografía
Septa caps are not processsed, pre-cleaned, nor certified.
Pack or case of vial closures do not contain any documentation, such as Certificate of Analysis or Cleaning Protocol Form. Septa caps are designed for use with C&G glass vials, all of which use a 24-414 closure.
Septa caps are polyprolylene open-top caps with a PTFE septa ultrasonically bonded (not glued). These are one-piece caps. The PTFE septa is a 3.1mm fluoropolymer resin/silicone septa laminate (overall thickness 0.125", ±0.006", silicone thickness is 0.115", PTFE is 0.010").