VWR® Boston Round Bottles, Narrow Mouth, Clear and Amber
Proveedor: VWR
VWR®
293078 292989 293049 292997 293075 292942 292933 293042 293054 293005 293014 292926 292922 292996 292947 292999 299897 292977 293074 292960 293057 293069 292991 293052 293072 293041 292967 293038 292971 293013 293067 293083 292916 292938 293000 293064 292985 293019 VWR89042-640 293044 292994 VWR89042-634 293062 293017 293074 292958 293016 292988 292986 292998 292992 292918 293004 293076 293023 292995 293079 VWR89042-710 292987 293027 292929 293047 292993 293002 293082 293010 293008 292944 293003 293060
10861-866CS 1288.29 MXN
10861-820 10861-788 10861-866 10861-860 10861-862 10862-504 10862-508 10862-510 10861-816 10861-818 10861-812 10861-856 10861-858 10861-814 10861-874 10861-798 76378-340 10861-754 10861-876 10861-998 10861-793 10861-870 10861-872 10861-796 10861-752 10861-790 76419-674 10862-502 10862-500 10861-948 10861-944 10861-900 10861-868 10861-946 10861-764 10861-886 10861-800 10861-888 10861-767 10861-881 10861-760 10861-882 10861-762 10861-884 10862-498 10861-916 10862-490 10861-758 10861-914 10862-492 10861-852 10861-896 10861-776 10861-898 10861-810 10861-778 10861-894 10861-850 10861-774 10862-514 10861-770 10862-000 10861-806 10861-808 10861-768 10861-922 10861-802 76329-568 10861-804 10861-848
VWR® Boston Round Bottles, Narrow Mouth, Clear and Amber
Frascos Botellas de vidrio
VWR® Brand Boston Round Bottles are made from soda-lime glass that conforms to USP Type III requirements.
  • Clear or amber coloring
  • Available in a variety of pack sizes
  • Narrow mouth
Bottles are available in clear or amber formats.
Several packaging options are offered for this line of Boston Round Bottles. Bottles are available in a convience pack with closures attached to help maintain cleanliness, they are available in a bulk pack with unattached caps in bags, they are available in a bulk pack without caps, or they are available in a bulk pack with caps attached.