VWR® Packer Style Bottles, Amber, Wide Mouth
Proveedor: VWR
VWR®
293165 293174 293138 293162 293089 293150 293112 293170 293125 293111 293164 293158 293153 293151 293132 293131 293139 293140 293154 293163 293115 293119 293175 293094 293159 293085 293106 293183 293093 293182 293124 293088 293141 293172 293176
10861-786CS 1180.55 MXN
10861-786 10861-842 10861-844 10861-782 10861-784 10861-960 10861-840 10862-008 10862-014 10862-012 10862-018 10862-016 10861-838 10862-010 10861-956 10861-836 10861-958 10861-830 10861-952 10861-930 10861-832 10861-954 10861-950 10862-006 10861-908 10862-004 10861-926 10861-904 10861-828 10861-928 10861-906 10862-020 10861-846 10861-902 10861-826
VWR® Packer Style Bottles, Amber, Wide Mouth
Frascos Botellas de vidrio
VWR® Amber wide mouth packer style bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • Variety of ordering options
  • Wide mouth allows for easy dispensation of contents
Bottles are available in three packaging options. They may be purchased in convenience packs with closures attached, in bulk packs with unattached closures in separate bags, or in bulk packs without closures.