VWR® French Square Bottles, Clear
Proveedor: VWR
VWR®
VWR89043-740 293037 VWR89043-732 293040 293024 293035 293018 293028 293050 VWR89043-670 293026 293015 297678 293036 293032 293022 293020 293033 293030 VWR89043-734 VWR89043-738 VWR89043-736 293034 293053 293012 293009 293039 293048 293045 293029 293006 293043
10862-198CS 2042.46 MXN
76404-282 10862-206 10862-108 10862-156 10862-110 10862-198 10862-154 10862-138 10862-136 10862-192 10862-094 10862-098 10862-194 10862-150 10862-096 10862-200 10862-146 10862-102 10862-088 10862-144 10862-100 10862-342 10862-188 10862-204 10862-106 10862-202 10862-148 10862-182 10862-180 10862-086 10862-140 10862-184
VWR® French Square Bottles, Clear
Frascos Botellas de vidrio
VWR® clear french square style bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
Three packaging options are available for these VWR® french square bottles. They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures included in separate bags, or bulk packed without closures.
Envase: Available in a variety of package sizes with different closure. options