VWR® Straight Sided Jars, Clear and Amber
Proveedor: VWR
VWR®
293211 293147 293152 293189 VWR89043-286 293205 293192 293188 293207 293196 293200 293203 293191 293185 293208 293199 293161 293193 293212 293194 293202 293209 293149 293213 293186 293198 293178 293206 293184 293204 293187 293181 293190 293197 293195 327112 293210 293148 293148 293160
10861-964CS 1052.99 MXN
10861-962 10861-940 10861-964 10861-942 76329-560 10862-112 10862-036 10862-078 10862-056 10862-059 10862-114 10862-038 10862-070 10862-076 10862-054 10861-934 10862-030 10862-074 10861-996 10861-932 76329-572 76329-550 10861-992 10861-994 10861-972 10862-024 10862-046 10862-068 76329-628 10862-124 10862-122 77394-648 10862-028 10862-128 10862-048 76329-566 10862-060 10862-080 10861-966 10861-968
VWR® Straight Sided Jars, Clear and Amber
Tarros Tarros para uso general
VWR® Straight Sided Jars are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • Clear and amber options
  • Straight sides, without shoulders for maximum storage capacity
Jars are available in multiple packaging options.