mySPIN™ Mini Centrifuges Series, Thermo Scientific
Catalog # 10199-452
Supplier: THERMO FISHER SCIENTIFIC MX
Specifications
- Description:mySPIN 6 Mini Centrifuge
- Cat. no.:10199-452
- W×D×H:5 x 6 x 4.1''
- Electrical:110/240 VAC
- Maximum Speed:6,000 rpm
- Maximum RCF:2000×g
- Maximum Capacity:6× 2 ml
- Weight:0.74 kg (1.6 lbs.)
- Supplier No.:75004061
Specifications
About this item
Bring increased efficiency and user comfort to your lab with mySPIN™ 6 and 12 mini centrifuges.
- Quiet operation, low vibration
- Ability to perform fast spins with higher RCF requirements
- Palm-shaped lid for left- or right-handed users
- mySPIN 12 centrifuge is ideal for most protocols requiring fast spins and appropriate for PCR, microfilter cell separation, and HPLC protocols
- mySPIN 6 is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL)
Tool-free rotors and quick-spin options make them fast and easy to use. Benefit from a compact footprint that fits into your workstation and ergnonomic design that stands out. Ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.